Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c19orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126559.1 | NP_001120031.1 | 6 - 7 | 655 | 60 | |
XM_012967985.1 | XP_012823439.1 | 6 - 7 | 729 | 60 | ||
XM_012967986.2 | XP_012823440.1 | 6 - 7 | 718 | 60 | ||
XM_012967987.2 | XP_012823441.1 | 6 - 7 | 760 | 60 | ||
XM_012967988.2 | XP_012823442.1 | 6 - 7 | 724 | 60 | ||
XM_012967989.2 | XP_012823443.1 | 6 - 7 | 693 | 60 | ||
XM_018096154.1 | XP_017951643.1 | 6 - 7 | 689 | 60 | ||
GenBank mRNA | BC158337.1 | - | 6 - 7 | 655 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |