Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126677.1 | NP_001120149.1 | 13 - 14 | 1846 | 80 | |
XM_012956449.2 | XP_012811903.1 | 12 - 13 | 1831 | 80 | ||
GenBank mRNA | BC159142.1 | - | 13 - 14 | 1846 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |