Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126748.1 | NP_001120220.1 | 1 - 2 | 260 | 92 | |
GenBank mRNA | BC159399.1 | - | 1 - 2 | 260 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |