Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: carnmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126522.1 | NP_001119994.1 | 1 - 2 | 513 | 67 | |
GenBank mRNA | BC158255.1 | - | 1 - 2 | 513 | 67 | |
CU024918.1 | - | 1 - 2 | 138 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |