Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127982.1 | NP_001121454.1 | 4 - 5 | 2183 | 74 | |
XM_018096166.1 | XP_017951655.1 | 3 - 4 | 2043 | 74 | ||
GenBank mRNA | BC166213.1 | - | 4 - 5 | 2183 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |