Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112422.1 | NP_001105892.1 | 369 | 71 | ||
GenBank mRNA | U44087.1 | - | 369 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |