Search
Reduced background staining afforded by Image-iT® FX signal enhancer. Mouse brain cryosections were permeabilized and antigen retrieval was carried out. The sections were then treated for 30 minutes with Image-iT® FX signal enhancer (Cat. no. I36933, left) or left untreated (right). Sections were labeled with the neural cell body selective antibody anti–Hu C/D (Cat. no. A21271) and visualized using TSA Kit #2 (Cat. no. T20912) with the HRP conjugate of goat anti–mouse IgG and Alexa Fluor® 488 tyramide. Sections were mounted using the reagents in the ProLong® Antifade Kit (Cat. no. P7481).
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Phagocytosis in MMM Cells using pHrodo™ Red E. coli BioParticles® Conjugate® (Catalog # P35361) Go ›
Muntjac skin fibroblast labeled with rhodamine phalloidin. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Immunocytochemistry using Alpha-Tubulin Monoclonal Antibody, Mouse Go ›