Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your analyses needs.
View the relevant questions below:
Having problems with your experiment? Visit 
Troubleshooting
No. However, it is a good idea to calibrate all scan settings at least once a year. In this way, if the zoom scan setting is used in the future, it can be reasonably easy to calibrate at that time.
Scan ModeLCQ Fleet Scan Rate (amu/s)LTQ & LXQ Scan Rate (amu/s)LTQ Velos Scan Rate (amu/s)LTQ Velos Pro Scan Rate (amu/s)
RapidNANANA66.000
Normal12.50016.70033.30033.300
Enhanced5.0005.0005.0005.000
Zoom1.1001.1001.1001.100
Ultra ZoomNA272727
Turbo125.000125.000125.000125.000
Proper Cal Mix
NL
The tune file contains four items: Source parameters, ion optic parameters, AGC targets, and scan time settings.
Need more information? Contact us ›
CMD SchemaApp code