Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVSNHYFLLC VNLPLREIHT PGPSPSCLGD SLAETTLSED TTDSVGSASP HGSSEKSSSF SLSSTEVHMV RPGYSHRVSL PTSPRILATS PYPETDSAFF EPSHLTSAAD EGAVQVSRRT ISSNSFSPEV FVLPVDVEKE NAHFYVADMI ISAMEKMKCN ILSQQQTESW SKEVSGLLGS DQPDSEMTFD TNIKQESGSS TSSYSGYEGC AVLQVSPVTE TRTYHDVKEI CKCDVDEFVI LELGDFNDIT ETCSCSCSSS KSVTYEPDFN SAELLAKELY RVFQKCWILS VVNSQLAGSL SAAGSIVVNE ECVRKDFESS MNVVQEIKFK SRIRGTEDWA PPRFQIIFNI HPPLKRDLVV AAQNFFCAGC GTPVEPKFVK RLRYCEYLGK YFCDCCHSYA ESCIPARILM MWDFKKYYVS NFSKQLLDSI WHQPIFNLLS IGQSLYAKAK ELDRVKEIQE QLFHIKKLLK TCRFANSALK EFEQVPGHLT DELHLFSLED LVRIKKGLLA PLLKDILKAS LAHVAGCELC QGKGFICEFC QNTTVIFPFQ TATCRRCSAC RACFHKQCFQ SSECPRCARI TARRKLLESV ASAAT
This gene encodes a cysteine-rich protein that contains a putative zinc-RING and/or ribbon domain. The encoded protein is related to Run domain Beclin-1-interacting and cysteine-rich domain-containing protein, which plays a role in endocytic trafficking and autophagy. In cervical cancer cell lines, this gene is expressed at low levels and may function as a tumor suppressor. Promoter hypermethylation of this gene is observed in cervical cancer cell lines and tissue derived from human patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIAA0226-like; Pacer; Protein associated with UVRAG as autophagy enhancer; Protein Rubicon-like; uncharacterized protein KIAA0226-like
Gene Aliases: C13orf18; Gm744; Gm911; KIAA0226L; RUBCNL
UniProt ID: (Human) Q9H714
Entrez Gene ID: (Mouse) 271221, (Human) 80183
Molecular Function: protein-binding activity modulator phosphatase inhibitor phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support