Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
1 µg/mL of MA5-45587 was sufficient for detection of TARP Gamma2/4/8 in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 35-55kDa. Does not cross-react with TARPGamma3.
This antibody was formerly sold as clone S245-36.
Voltage-dependent calcium channel gamma-2 subunit, TARPGamma2 or Stargazin is encoded by the gene CACNG2 and is the transmembrane gamma subunit of the L-type calcium channel. Stargazin is in involved in regulation and trafficking of the ligand gated ion channel AMPA receptors. It is thought to play a role in learning and memory. Diseases associated with this gene include forms of deafness, Mental Retardation and childhood absnce epilepsy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium channel, voltage-dependent, gamma subunit 2; calcium channel, voltage-dependent, gamma subunit 4; calcium channel, voltage-dependent, gamma subunit 8; Neuronal voltage-gated calcium channel gamma-2 subunit; Neuronal voltage-gated calcium channel gamma-4 subunit; Neuronal voltage-gated calcium channel gamma-8 subunit; Stargazin; TARP gamma 2; TARP gamma 8; TARP gamma-2; TARP gamma-4; TARP gamma-8; Transmembrane AMPAR regulatory protein gamma-2; Transmembrane AMPAR regulatory protein gamma-4; Transmembrane AMPAR regulatory protein gamma-8; Voltage-dependent calcium channel gamma-2 subunit; Voltage-dependent calcium channel gamma-4 subunit; Voltage-dependent calcium channel gamma-8 subunit
Gene Aliases: AI413107; AW060990; AW491861; B230105C07Rik; B930041E13Rik; CACNG2; CACNG4; CACNG6; CACNG8; Ipr328; mir-935; MRD10; stargazer; stargazin; Stg; wag; waggler
UniProt ID: (Human) Q9Y698, (Human) Q9UBN1, (Human) Q8WXS5, (Rat) Q71RJ2, (Mouse) O88602, (Rat) Q8VHW9, (Mouse) Q9JJV4, (Rat) Q8VHW5
Entrez Gene ID: (Human) 10369, (Human) 27092, (Human) 59283, (Rat) 84347, (Mouse) 12300, (Rat) 140725, (Mouse) 54377, (Rat) 140729, (Mouse) 81905
Molecular Function: voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support