Product References
A protocol for generating induced T cells by reprogramming B cells in vivo.
Cell regeneration (London, England)
Weng Q,Hu F,Zhang M,Dong Y,Lv C,Wang Y,Liu X,Wang J
MA5-17859 was used in Immunohistochemistry to describe a protocol for generating functional T cells from Hoxb5-expressing B cells in vivo.
Sat Sep 01 00:00:00 EDT 2018
Conventional bone marrow-derived dendritic cells contribute to toll-like receptor-independent production of alpha/beta interferon in response to inactivated parapoxvirus ovis.
Journal of virology
Siegemund S,Hartl A,von Buttlar H,Dautel F,Raue R,Freudenberg MA,Fejer G,Büttner M,Köhler G,Kirschning CJ,Sparwasser T,Alber G
MA5-17859 was used in flow cytometry to study the effect of inactivated Parapoxvirus ovis on dendritic cells
Tue Sep 01 00:00:00 EDT 2009
Role of T cells in innate and adaptive immunity against murine Burkholderia pseudomallei infection.
The Journal of infectious diseases
Haque A,Easton A,Smith D,O'Garra A,Van Rooijen N,Lertmemongkolchai G,Titball RW,Bancroft GJ
MA5-17859 was used in flow cytometry to study the in vivo role of cytokine- and antigen-mediated T cell activation in the resistance to Burkholderia pseudomallei
Wed Feb 01 00:00:00 EST 2006
TLR4, TLR9 and MyD88 are not required for the positive selection of autoreactive B cells into the primary repertoire.
European journal of immunology
Silver K,Ferry H,Crockford T,Cornall RJ
MA5-17859 was used in flow cytometry to demonstrate that the effects of Toll-like receptors on autoimmunity occur in the established repertoire and not during B cell development
Thu Jun 01 00:00:00 EDT 2006
Tick saliva inhibits differentiation, maturation and function of murine bone-marrow-derived dendritic cells.
Immunology
Cavassani KA,Aliberti JC,Dias AR,Silva JS,Ferreira BR
MA5-17859 was used in flow cytometry to assess the effects of Rhipicephalus sanguineus tick saliva on dendritic cell differentiation and maturation
Tue Feb 01 00:00:00 EST 2005
Time-course expression of Toll-like receptors 2 and 4 in inflammatory bowel disease and homeostatic effect of VIP.
Journal of leukocyte biology
Gomariz RP,Arranz A,Abad C,Torroba M,Martinez C,Rosignoli F,Garcia-Gómez M,Leceta J,Juarranz Y
MA5-17859 was used in flow cytometry to study TLR2 and 4 expression in a trinitrobenzene sulfonic acid-induced colitis model of Crohn's disease
Mon Aug 01 00:00:00 EDT 2005
Modulation of interferon-induced genes by lipoxin analogue in anti-glomerular basement membrane nephritis.
Journal of the American Society of Nephrology : JASN
Ohse T,Ota T,Kieran N,Godson C,Yamada K,Tanaka T,Fujita T,Nangaku M
MA5-17859 was used in flow cytometry to assess the effect of aspirin-triggered 15-epi-lipoxin A4 in experimental anti-glomerular basement membrane antibody nephritis
Thu Apr 01 00:00:00 EST 2004
Characterization of the mouse analogues of CD59 using novel monoclonal antibodies: tissue distribution and functional comparison.
Immunology
Harris CL,Hanna SM,Mizuno M,Holt DS,Marchbank KJ,Morgan BP
MA5-17859 was used in flow cytometry to determine the expression of CD59a and CD59b.
Thu May 01 00:00:00 EDT 2003
Macrophage C-type lectin on bone marrow-derived immature dendritic cells is involved in the internalization of glycosylated antigens.
Glycobiology
Denda-Nagai K,Kubota N,Tsuiji M,Kamata M,Irimura T
MA5-17859 was used in flow cytometry to study expression of a C-type lectin specific for galactose and N-acetylgalactosamine on murine dendritic cells.
Mon Jul 01 00:00:00 EDT 2002
Immunosuppression during acute Trypanosoma cruzi infection: involvement of Ly6G (Gr1(+))CD11b(+ )immature myeloid suppressor cells.
International immunology
Goñi O,Alcaide P,Fresno M
MA5-17859 was used in flow cytometry to elucidate the immunosuppressive mechanism that occurs during T. cruzi infection.
Tue Oct 01 00:00:00 EDT 2002
Candida albicans infection enhances immunosuppression induced by cyclophosphamide by selective priming of suppressive myeloid progenitors for NO production.
Cellular immunology
Angulo I,Jiménez-Díaz MB,García-Bustos JF,Gargallo D,de las Heras FG,Muñoz-Fernández MA,Fresno M
MA5-17859 was used in flow cytometry to examine the T cell responses in cyclophosphamide-treated mice after systemic infection with Candida albicans.
Wed Feb 05 00:00:00 EST 2003
Nitric oxide-producing CD11b(+)Ly-6G(Gr-1)(+)CD31(ER-MP12)(+) cells in the spleen of cyclophosphamide-treated mice: implications for T-cell responses in immunosuppressed mice.
Blood
Angulo I,de las Heras FG,García-Bustos JF,Gargallo D,Muñoz-Fernández MA,Fresno M
MA5-17859 was used in flow cytometry to study T-cell unresponsiveness in cyclophosphamide treated mice.
Sat Jan 01 00:00:00 EST 2000
Identification of an early T cell progenitor for a pathway of T cell maturation in the bone marrow.
Proceedings of the National Academy of Sciences of the United States of America
Dejbakhsh-Jones S,Strober S
MA5-17859 was used in flow cytometry to identify a rare population of cells in the adult mouse bone marrow that generates CD4(+) and CD8(+) TCRalphabeta(+) T cells.
Tue Dec 07 00:00:00 EST 1999
Immunophenotypical and functional heterogeneity of dendritic cells generated from murine bone marrow cultured with different cytokine combinations: implications for anti-tumoral cell therapy.
Immunology
Masurier C,Pioche-Durieu C,Colombo BM,Lacave R,Lemoine FM,Klatzmann D,Guigon M
MA5-17859 was used in flow cytometry to characterize in vitro-generated dendritic cells.
Thu Apr 01 00:00:00 EST 1999
Cellular and molecular characterization of the scurfy mouse mutant.
Journal of immunology (Baltimore, Md. : 1950)
Clark LB,Appleby MW,Brunkow ME,Wilkinson JE,Ziegler SF,Ramsdell F
MA5-17859 was used in flow cytometry to further characterize mice hemizygous for scurfy.
Mon Mar 01 00:00:00 EST 1999
Three populations of mouse lymph node dendritic cells with different origins and dynamics.
Journal of immunology (Baltimore, Md. : 1950)
Salomon B,Cohen JL,Masurier C,Klatzmann D
MA5-17859 was used in flow cytometry to characterize different populations of mouse lymph node dendritic cells.
Thu Jan 15 00:00:00 EST 1998
The preclinical pharmacological profile of the potent and selective leukotriene B4 antagonist CP-195543.
The Journal of pharmacology and experimental therapeutics
Showell HJ,Conklyn MJ,Alpert R,Hingorani GP,Wright KF,Smith MA,Stam E,Salter ED,Scampoli DN,Meltzer S,Reiter LA,Koch K,Piscopio AD,Cortina SR,Lopez-Anaya A,Pettipher ER,Milici AJ,Griffiths RJ
MA5-17859 was used in flow cytometry to characterize a selective and potent leukotriene B4 receptor antagonist.
Mon Jun 01 00:00:00 EDT 1998
Immunologic memory induced by a glycoconjugate vaccine in a murine adoptive lymphocyte transfer model.
Infection and immunity
Guttormsen HK,Wetzler LM,Finberg RW,Kasper DL
MA5-17859 was used in flow cytometry to develop a mouse model to study the ability of a glycoprotein conjugate vaccine to induce immunologic memory for the polysaccharide moiety.
Fri May 01 00:00:00 EDT 1998
Mechanisms of allografted tumor rejection: the roles of T cells in allograft rejection mediated by a type of bone marrow-derived macrophage.
Microbiology and immunology
Ushio-Umeda Y,Yoshida R
MA5-17859 was used in flow cytometry to elucidate the roles of T cells in the AIM-mediated rejection of tumor cells.
Thu Apr 02 00:00:00 EST 1998
Thy.1lowCD3- cells sorted from nylon wool-passed bone marrow cells can augment the H-2 identical but not non-identical cytotoxic T lymphocyte precursors in mixed lymphocyte cultures.
Microbiology and immunology
Mukouyama H,Tanabe MJ
MA5-17859 was used in flow cytometry to characterize T cells obtained from nylon wool-passed murine bone marrow.
Tue May 23 00:00:00 EDT 1995
CD3+ and CD4+ cells adoptively transfer experimental hypersensitivity pneumonitis.
The American review of respiratory disease
Schuyler M,Gott K,Shopp G,Crooks L
MA5-17859 was used in flow cytometry to describe the cells that transfer adoptive murine experimental hypersensitivity pneumonitis.
Tue Dec 01 00:00:00 EST 1992
Mononuclear phagocytes: a major population of effector cells responsible for rejection of allografted tumor cells in mice.
Proceedings of the National Academy of Sciences of the United States of America
Yoshida R,Takikawa O,Oku T,Habara-Ohkubo A
MA5-17859 was used in flow cytometry to examine the cells that infiltrated into the rejection site of allografted tumors.
Fri Feb 15 00:00:00 EST 1991