Product References
STS1 and STS2 Phosphatase Inhibitor Baicalein Enhances the Expansion of Hematopoietic and Progenitor Stem Cells and Alleviates 5-Fluorouracil-Induced Myelosuppression.
International journal of molecular sciences
Li N,Wang Y,Wang A,Zhang J,Jia C,Yu C,Song Z,Wang S,Liu L,Yi J,Bao Y,Huang Y,Sun L
Published figure using CD135 (Flt3) monoclonal antibody (Product # 15-1351-82) in Flow Cytometry
Fri Feb 03 00:00:00 EST 2023
Protocol for isolation and analysis of the leukemia stem cells in BCR-ABL-driven chronic myelogenous leukemia mice.
STAR protocols
Liu C,Lin X,Jin Y,Pan J
15-1351-82 was used in flow cytometry to describe practical procedures for sorting and analysis of leukemia stem cells (LSCs) to improve our understanding of chronic myelogenous leukemia (CML).
Fri Mar 17 00:00:00 EDT 2023
Erythropoietin directly remodels the clonal composition of murine hematopoietic multipotent progenitor cells.
eLife
Eisele AS,Cosgrove J,Magniez A,Tubeuf E,Tenreira Bento S,Conrad C,Cayrac F,Tak T,Lyne AM,Urbanus J,Perié L
15-1351-82 was used in flow cytometry to show that EPO does act directly on MPP independent of the niche and modulates fate by remodeling the clonal composition of the MPP pool.
Tue Feb 15 00:00:00 EST 2022
Aged hematopoietic stem cells are refractory to bloodborne systemic rejuvenation interventions.
The Journal of experimental medicine
Ho TT,Dellorusso PV,Verovskaya EV,Bakker ST,Flach J,Smith LK,Ventura PB,Lansinger OM,Hérault A,Zhang SY,Kang YA,Mitchell CA,Villeda SA,Passegué E
Published figure using CD135 (Flt3) monoclonal antibody (Product # 15-1351-82) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
TNF-α-induced alterations in stromal progenitors enhance leukemic stem cell growth via CXCR2 signaling.
Cell reports
Agarwal P,Li H,Choi K,Hueneman K,He J,Welner RS,Starczynowski DT,Bhatia R
15-1351-82 was used in Flow Cytometry to find that TNF-α-mediated alterations in CML BM stromal niches enhance support of LSC maintenance and growth via CXCL1-CXCR2 signaling and that CXCR2 inhibition effectively depletes CML LSCs.
Tue Jul 13 00:00:00 EDT 2021
Mesenchymal stem cells attenuate liver fibrosis by targeting Ly6Chi/lo macrophages through activating the cytokine-paracrine and apoptotic pathways.
Cell death discovery
Li YH,Shen S,Shao T,Jin MT,Fan DD,Lin AF,Xiang LX,Shao JZ
15-1351-82 was used in Flow Cytometry to provide insights into the functional role of bone marrow-derived sesenchymal stem cell in alleviating liver fibrosis.
Mon Sep 13 00:00:00 EDT 2021
Adaptive response to inflammation contributes to sustained myelopoiesis and confers a competitive advantage in myelodysplastic syndrome HSCs.
Nature immunology
Muto T,Walker CS,Choi K,Hueneman K,Smith MA,Gul Z,Garcia-Manero G,Ma A,Zheng Y,Starczynowski DT
15-1351 was used in Flow cytometry/Cell sorting to show that chronic inflammation is a determinant for the competitive advantage of myelodysplastic syndrome (MDS) hematopoietic stem and progenitor cells (HSPCs) and for disease progression.
Fri May 01 00:00:00 EDT 2020
Deregulated Notch and Wnt signaling activates early-stage myeloid regeneration pathways in leukemia.
The Journal of experimental medicine
Kang YA,Pietras EM,Passegué E
Published figure using CD135 (Flt3) monoclonal antibody (Product # 15-1351-82) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2020
Zinc finger and BTB domain-containing protein 46 is essential for survival and proliferation of acute myeloid leukemia cell line but dispensable for normal hematopoiesis.
Chinese medical journal
Liu YY,Xiao FF,Yang BJ,Li X,Xu SN,Chen ZW,Li P,Huang YX,Fu XM,Huang XQ,Zheng GL,Chen JP,Hou Y
Published figure using CD135 (Flt3) monoclonal antibody (Product # 15-1351-82) in Flow Cytometry
Mon Jul 20 00:00:00 EDT 2020
Connecting secretome to hematopoietic stem cell phenotype shifts in an engineered bone marrow niche.
Integrative biology : quantitative biosciences from nano to macro
Gilchrist AE,Harley BAC
15-1351-82 was used in Flow Cytometry to investigate the mechanism of how marrow-derived mesenchymal stromal and progenitor cells impacts hematopoietic stem cells fate decisions.
Fri Jul 10 00:00:00 EDT 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD135 (Flt3) monoclonal antibody (Product # 15-1351-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
An Id2RFP-Reporter Mouse Redefines Innate Lymphoid Cell Precursor Potentials.
Immunity
Xu W,Cherrier DE,Chea S,Vosshenrich C,Serafini N,Petit M,Liu P,Golub R,Di Santo JP
Published figure using CD135 (Flt3) monoclonal antibody (Product # 15-1351-82) in Flow Cytometry
Tue Apr 16 00:00:00 EDT 2019
Polychromic Reporter Mice Reveal Unappreciated Innate Lymphoid Cell Progenitor Heterogeneity and Elusive ILC3 Progenitors in Bone Marrow.
Immunity
Walker JA,Clark PA,Crisp A,Barlow JL,Szeto A,Ferreira ACF,Rana BMJ,Jolin HE,Rodriguez-Rodriguez N,Sivasubramaniam M,Pannell R,Cruickshank J,Daly M,Haim-Vilmovsky L,Teichmann SA,McKenzie ANJ
Published figure using CD135 (Flt3) monoclonal antibody (Product # 15-1351-82) in Flow Cytometry
Tue Jul 16 00:00:00 EDT 2019
Mesenchymal Niche-Specific Expression of Cxcl12 Controls Quiescence of Treatment-Resistant Leukemia Stem Cells.
Cell stem cell
Agarwal P,Isringhausen S,Li H,Paterson AJ,He J,Gomariz Á,Nagasawa T,Nombela-Arrieta C,Bhatia R
15-1351-82 was used in Flow Cytometry to highlight a critical role of niche-specific effects of Cxcl12 expression in maintaining quiescence of TKI-resistant LSC populations.
Thu May 02 00:00:00 EDT 2019
MicroRNA-155 Protects Group 2 Innate Lymphoid Cells From Apoptosis to Promote Type-2 Immunity.
Frontiers in immunology
Knolle MD,Chin SB,Rana BMJ,Englezakis A,Nakagawa R,Fallon PG,Git A,McKenzie ANJ
15-1351 was used in Flow cytometry/Cell sorting to identify a subset of micro-RNAs that are regulated upon group-2 innate lymphoid cells (ILC2) activation, and establish a specific role for miR-155 in regulating ILC2 survival following activation.
Mon Sep 30 00:00:00 EDT 2019
Ubiquitination of hnRNPA1 by TRAF6 links chronic innate immune signaling with myelodysplasia.
Nature immunology
Fang J,Bolanos LC,Choi K,Liu X,Christie S,Akunuru S,Kumar R,Wang D,Chen X,Greis KD,Stoilov P,Filippi MD,Maciejewski JP,Garcia-Manero G,Weirauch MT,Salomonis N,Geiger H,Zheng Y,Starczynowski DT
15-1351 was used in Flow cytometry/Cell sorting to examine the significance of ubiquitin signalling in coordinating RNA processing by Toll-like receptor pathways during an immune response and in premalignant haematologic diseases.
Wed Feb 01 00:00:00 EST 2017
LCN2 overexpression in bone enhances the hematopoietic compartment via modulation of the bone marrow microenvironment.
Journal of cellular physiology
Costa D,Principi E,Lazzarini E,Descalzi F,Cancedda R,Castagnola P,Tavella S
15135182 was used in flow cytometry to examine the function of lipocalin-2 in bone tissue
Wed Nov 01 00:00:00 EDT 2017
A single-cell resolution map of mouse hematopoietic stem and progenitor cell differentiation.
Blood
Nestorowa S,Hamey FK,Pijuan Sala B,Diamanti E,Shepherd M,Laurenti E,Wilson NK,Kent DG,Göttgens B
15-1351 was used in Flow cytometry/Cell sorting to report the development of an intuitive Web interface as a new community resource to permit visualisation of gene expression in haematopoietic stem and progenitor cells at single-cell resolution for any gene of choice.
Thu Aug 25 00:00:00 EDT 2016
Clec11a/osteolectin is an osteogenic growth factor that promotes the maintenance of the adult skeleton.
eLife
Yue R,Shen B,Morrison SJ
15-1351 was used in Flow cytometry/Cell sorting to study how Clec11a/Osteolectin maintains the adult skeleton by promoting the differentiation of mesenchymal progenitors into mature osteoblasts.
Tue Dec 13 00:00:00 EST 2016
Index sorting resolves heterogeneous murine hematopoietic stem cell populations.
Experimental hematology
Schulte R,Wilson NK,Prick JC,Cossetti C,Maj MK,Gottgens B,Kent DG
15-1351 was used in Flow cytometry/Cell sorting to indicate that when used in combination with single-cell functional assays, index sorting is a powerful tool for refining cell isolation strategies.
Tue Sep 01 00:00:00 EDT 2015
Oestrogen increases haematopoietic stem-cell self-renewal in females and during pregnancy.
Nature
Nakada D,Oguro H,Levi BP,Ryan N,Kitano A,Saitoh Y,Takeichi M,Wendt GR,Morrison SJ
15-1351 was used in Flow cytometry/Cell sorting to assess the effect of oestradiol administration on division of haematopoietic stem cells in males and females.
Thu Jan 23 00:00:00 EST 2014
HIF prolyl hydroxylase 2 (PHD2) is a critical regulator of hematopoietic stem cell maintenance during steady-state and stress.
Blood
Singh RP,Franke K,Kalucka J,Mamlouk S,Muschter A,Gembarska A,Grinenko T,Willam C,Naumann R,Anastassiadis K,Stewart AF,Bornstein S,Chavakis T,Breier G,Waskow C,Wielockx B
15-1351 was used in Flow cytometry/Cell sorting to study the role of PHD2 in HSC compartment maintenance.
Thu Jun 27 00:00:00 EDT 2013
An information theoretic, microfluidic-based single cell analysis permits identification of subpopulations among putatively homogeneous stem cells.
PloS one
Glotzbach JP,Januszyk M,Vial IN,Wong VW,Gelbard A,Kalisky T,Thangarajah H,Longaker MT,Quake SR,Chu G,Gurtner GC
15-1351 was used in Flow cytometry/Cell sorting to develop a novel method to characterize and predict transcriptional programs across individual homogeneous stem cells.
Wed Nov 02 00:00:00 EDT 2011
CX3CR1+ CD115+ CD135+ common macrophage/DC precursors and the role of CX3CR1 in their response to inflammation.
The Journal of experimental medicine
Auffray C,Fogg DK,Narni-Mancinelli E,Senechal B,Trouillet C,Saederup N,Leemput J,Bigot K,Campisi L,Abitbol M,Molina T,Charo I,Hume DA,Cumano A,Lauvau G,Geissmann F
15-1351 was used in Flow cytometry/Cell sorting to show that macrophage/DC precursors give rise to conventional DCs,.
Mon Mar 16 00:00:00 EDT 2009
Niche recycling through division-independent egress of hematopoietic stem cells.
The Journal of experimental medicine
Bhattacharya D,Czechowicz A,Ooi AG,Rossi DJ,Bryder D,Weissman IL
15-1351 was used in Flow cytometry/Cell sorting to provide insight as to how HSC replacement can occur despite the residence of endogenous HSCs in niches.
Mon Nov 23 00:00:00 EST 2009