Product References
Frequent aerogenous spread with decreased E-cadherin expression of ROS1-rearranged lung cancer predicts poor disease-free survival.
Lung cancer (Amsterdam, Netherlands)
Jin Y,Sun PL,Park SY,Kim H,Park E,Kim G,Cho S,Kim K,Lee CT,Chung JH
MA5-14408 was used in immunohistochemistry to characterize ROS1-rearranged tumors
Tue Sep 01 00:00:00 EDT 2015
Comparison of the clinicopathological features of invasive ductal, invasive lobular, and mixed (invasive ductal + invasive lobular) carcinoma of the breast.
Breast cancer (Tokyo, Japan)
Zengel B,Yararbas U,Duran A,Uslu A,Elıyatkın N,Demırkıran MA,Cengiz F,Şimşek C,Postacı H,Vardar E,Durusoy R
MA5-14408 was used in immunohistochemistry to perform a retrospective study of the clinicopathology of various forms of invasive breast carcinoma
Wed Jul 01 00:00:00 EDT 2015
Immunohistochemical expression of CD44 standard and E-cadherin in atypical leiomyoma and leiomyosarcoma of the uterus.
Journal of obstetrics and gynaecology : the journal of the Institute of Obstetrics and Gynaecology
Kaygusuz EI
MA5-14408 was used in immunohistochemistry - paraffin section to assess the diagnostic role of CD44 and E-cadherin in atypical leiomyoma and leiomyosarcoma.
Wed Apr 01 00:00:00 EDT 2015
The effects of sonic hedgehog signaling pathway components on non-small-cell lung cancer progression and clinical outcome.
World journal of surgical oncology
Hwang J,Kang MH,Yoo YA,Quan YH,Kim HK,Oh SC,Choi YH
MA5-14408 was used in immunohistochemistry - paraffin section to study non-small-cell lung cancer progression and clinical outcome by the effects of sonic hedgehog signaling pathway components
Thu Aug 21 00:00:00 EDT 2014
A comprehensive comparative analysis of the histomorphological features of ALK-rearranged lung adenocarcinoma based on driver oncogene mutations: frequent expression of epithelial-mesenchymal transition markers than other genotype.
PloS one
Kim H,Jang SJ,Chung DH,Yoo SB,Sun P,Jin Y,Nam KH,Paik JH,Chung JH
MA5-14408 was used in immunohistochemistry to perform a detailed comparative analysis of ALK-rearranged and ALK-negative lung adenocarcinomas
Tue Feb 24 00:00:00 EST 2015
Epithelial-mesenchymal transition in breast cancer correlates with high histological grade and triple-negative phenotype.
Histopathology
Jeong H,Ryu YJ,An J,Lee Y,Kim A
MA5-14408 was used in immunohistochemistry to study the correlation of epithelial-mesenchymal transition with triple negative phenotype and high histological grade in breast cancer
Tue May 01 00:00:00 EDT 2012
Apoptosis in the developing human heart resembles apoptosis in epithelial tissues.
Cell and tissue research
Jerše M,Zidar N
MA5-14408 was used in immunohistochemistry to investigate the process of apoptosis in human heart during development
Tue Mar 01 00:00:00 EST 2011
Down-regulation of microRNAs of the miR-200 family and miR-205, and an altered expression of classic and desmosomal cadherins in spindle cell carcinoma of the head and neck--hallmark of epithelial-mesenchymal transition.
Human pathology
Zidar N,Boštjančič E,Gale N,Kojc N,Poljak M,Glavač D,Cardesa A
MA5-14408 was used in immunohistochemistry to study the downregulation of specific microRNAs and altered expression of cadherins in spindle cell carcinoma of the head and neck
Fri Apr 01 00:00:00 EDT 2011
GLI2-mediated melanoma invasion and metastasis.
Journal of the National Cancer Institute
Alexaki VI,Javelaud D,Van Kempen LC,Mohammad KS,Dennler S,Luciani F,Hoek KS,Juàrez P,Goydos JS,Fournier PJ,Sibon C,Bertolotto C,Verrecchia F,Saule S,Delmas V,Ballotti R,Larue L,Saiag P,Guise TA,Mauviel A
MA5-14408 was used in immunohistochemistry to study the role of the Hedgehog pathway mediator GLI2 in regulating melanoma invasion and metastasis
Wed Aug 04 00:00:00 EDT 2010
An immunohistochemical study of colon adenomas and carcinomas: E-cadherin, Syndecan-1, Ets-1.
Pathology oncology research : POR
Pap Z,Pávai Z,Dénes L,Kovalszky I,Jung J
MA5-14408 was used in immunohistochemistry to investigate the immunohistochemical differences between colon adenomas and carcinomas
Tue Dec 01 00:00:00 EST 2009
Cadherin-catenin complex and transcription factor Snail-1 in spindle cell carcinoma of the head and neck.
Virchows Archiv : an international journal of pathology
Zidar N,Gale N,Kojc N,Volavsek M,Cardesa A,Alos L,Höfler H,Blechschmidt K,Becker KF
MA5-14408 was used in immunohistochemistry to examine the role of transcription factor Snail-1 in spindle cell carcinoma of the head and neck
Mon Sep 01 00:00:00 EDT 2008