Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAVAAVAAR RRRSWASLVL AFLGVCLGIT LAVDRSNFKT CEESSFCKRQ RSIRPGLSPY RALLDSLQLG PDSLTVHLIH EVTKVLLVLE LQGLQKNMTR FRIDELEPRR PRYRVPDVLV ADPPIARLSV SGRDENSVEL TMAEGPYKII LTARPFRLDL LEDRSLLLSV NARGLLEFEH QRAPRVSQGS KDPAEGDGAQ PEETPRDGDK PEETQGKAEK DEPGAWEETF KTHSDSKPYG PMSVGLDFSL PGMEHVYGIP EHADNLRLKV TEGGEPYRLY NLDVFQYELY NPMALYGSVP VLLAHNPHRD LGIFWLNAAE TWVDISSNTA GKTLFGKMMD YLQGSGETPQ TDVRWMSETG IIDVFLLLGP SISDVFRQYA SLTGTQALPP LFSLGYHQSR WNYRDEADVL EVDQGFDDHN LPCDVIWLDI EHADGKRYFT WDPSRFPQPR TMLERLASKR RKLVAIVDPH IKVDSGYRVH EELRNLGLYV KTRDGSDYEG WCWPGSAGYP DFTNPTMRAW WANMFSYDNY EGSAPNLFVW NDMNEPSVFN GPEVTMLKDA QHYGGWEHRD VHNIYGLYVH MATADGLRQR SGGMERPFVL ARAFFAGSQR FGAVWTGDNT AEWDHLKISI PMCLSLGLVG LSFCGADVGG FFKNPEPELL VRWYQMGAYQ PFFRAHAHLD TGRREPWLLP SQHNDIIRDA LGQRYSLLPF WYTLLYQAHR EGIPVMRPLW VQYPQDVTTF NIDDQYLLGD ALLVHPVSDS GAHGVQVYLP GQGEVWYDIQ SYQKHHGPQT LYLPVTLSSI PVFQRGGTIV PRWMRVRRSS ECMKDDPITL FVALSPQGTA QGELFLDDGH TFNYQTRQEF LLRRFSFSGN TLVSSSADPE GHFETPIWIE RVVIIGAGKP AAVVLQTKGS PESRLSFQHD PETSVLVLRK PGINVASDWS IHLR
This gene encodes the alpha subunit of glucosidase II and a member of the glycosyl hydrolase 31 family of proteins. The heterodimeric enzyme glucosidase II plays a role in protein folding and quality control by cleaving glucose residues from immature glycoproteins in the endoplasmic reticulum. Expression of the encoded protein is elevated in lung tumor tissue and in response to UV irradiation. Mutations in this gene cause autosomal-dominant polycystic kidney and liver disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-glucosidase 2; Glucosidase II subunit alpha; Neutral alpha-glucosidase AB
Gene Aliases: G2AN; GANAB; GIIA; GLUII; KIAA0088
UniProt ID: (Human) Q14697
Entrez Gene ID: (Human) 23193
Molecular Function: glucosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support