Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
1 µg/mL of MA5-45476 was sufficient for detection of TrpC7 in 10 µg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.| Detects approximately 100kDa.
TRPC7 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC7 is probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. TRPC7 is also activated by diacylglycerol (DAG). Further, TRPC7 may also be activated by intracellular calcium store depletion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: capacitative calcium channel TRPC7; hTRP7; likley ortholog of mouse transient receptor potential cation channel, subfamily C, member 7; mTRP7; putative capacitative calcium channel; Short transient receptor potential channel 7; transient receptor potential cation channel, subfamily C, member 7; Transient receptor protein 7; TRP-7; TRP7 protein; TrpC7
Gene Aliases: TRP7; TRPC7; Trrp8
UniProt ID: (Human) Q9HCX4, (Mouse) Q9WVC5
Entrez Gene ID: (Human) 57113, (Rat) 282822, (Mouse) 26946
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support