+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
ImmunoCAP Rapid Point-of-Care 
Delivering Reliable Results to Help Clinicians Determine the Course of Care
IMMUNOCAP™ RAPID FOR POINT-OF-CARE TESTING
Delivering Reliable Results to Help Clinicians Determine the Course of Care
ImmunoCAP Rapid is a handheld, self-contained unit that generates an IgE profile within minutes for adults and children with symptoms of wheezing and rhinitis. Just add capillary or whole blood into the sample well, wait 20 minutes, and read the result as a colored band. For patients who test positive to one or more allergens, clinicians can include these results in their diagnostic conclusion and initiate appropriate treatment quickly. But for those with negative results you can discuss treatment and further action—before the patient leaves your office or facility.
ImmunoCAP Rapid does not require a Phadia™ Laboratory System. It is a CE-marked product and also available in Japan.
High Positive Predictive Ability to Identify Triggering Allergens
ImmunoCAP Rapid is a point-of-care test for in vitro qualitative determination of allergen-specific IgE in human capillary blood or heparinized whole venous blood. In clinical studies, more than 95% of allergic patients studied were identified with ImmunoCAP Rapid test.1-3
ImmunoCAP Rapid offers a comprehensive solution from capillary blood sampling to the evaluation of test results, including all necessary equipment such as Blood Sampling Device, Assay Device and Developer Solution in each package. The Assay Device, with a symptom-oriented allergen profile, is a single use, disposable test device. It contains all the allergens to be tested and the necessary reagents for the reaction to occur. Ten allergens are included in each Assay Device. Results are read visually within 20 minutes. A pink-red line next to the allergen code indicates presence of specific IgE antibodies to that particular allergen.
ImmunoCAP Rapid gives results directly – clear and simple. ImmunoCAP Rapid is not available in all countries.
ImmunoCAP Rapid
Contact
Is your laboratory interested in expanding your range of testing tools?
If your laboratory has been considering upgrading or adding to your offerings, Thermo Fisher Scientific can help you find the right options for your needs.
References
  1. Hedlin G, Moreno C, Petersson CJ, Lilja G, Toledano FL, Garcia AN, Nordvall L, Palmqvist M, Rak S, Ahlstedt S, Borres MP: Allergy Diagnosis in Children and Adults: Performance of a New Point-of-Care Device, ImmunoCAP Rapid. World Allergy Organ J. 2009, 2: 138-43. 
  2. Diaz-Vazquez C, et al. Accuracy of ImmunoCAP Rapid in the diagnosis of allergic sensitization in children between 1 and 14 years with recurrent wheezing: The IReNE study. Pediatr Allergy Immunol 2009;20:601–609.
  3. Eigenmann PA, et al. The ImmunoCAP Rapid Wheeze/Rhinitis Child test is useful in the initial allergy diagnosis of children with respiratory symptoms. Pediatr Allergy Immunol 2009;20:772–779.