Software to support your laboratory's needs
PhadiaTM Information Data Manager (IDM) software helps your laboratory make the most of your Phadia Laboratory Systems. Not only can Phadia IDM manage assay runs, test requests and results, but it also can help your laboratory ensure quality control of data and inventory.
Phadia IDM, has all the key functionality required to operate Phadia Laboratory Systems. This software gives you the ability to seamlessly integrate up to 5 Phadia 100 instruments, 5 Phadia 250 instruments and 5 Phadia 1000 instruments in a single network.* In addition, Phadia IDM automates communication with other laboratory systems, such as Laboratory Information System (LIS) and Laboratory Automation System (LAS).
Offering several improvements to Phadia IDM, the new PhadiaTM Prime software provides better ease of use, new tests, test management and network connectivity.
*For laboratories using Phadia 2500 or Phadia 5000 instruments, only one instrument can be connected using Phadia IDM
If your laboratory has been considering a Phadia Laboratory System or upgrading from Phadia IDM to Phadia Prime we can help.