EDTA, RTU

Catalog number: 005501

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This EDTA solution is specifically formulated for use in our Heat Induced Epitope Retrieval (HIER) method. Supplied as a 100 mL aliquot of 20X concentrate or 1L ready-to-use format.

Specifications

Reagent Type: Antigen Retrieval Reagent
Product Size: 1 L
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products