Thermo Scientific Chemicals

Gallium(III) fluoride, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 013987.06
5 g, Each
Thermo Scientific Chemicals

Gallium(III) fluoride, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 013987.06
5 g, Each
Quantity
Catalog number: 013987.06
also known as 013987-06
Price (USD)
Price: 455.00
Online price: 386.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7783-51-9
Total Metal Impurities
0.001% max.

Description

Gallium(III) fluoride is employed in the preparation and study of optical fluorogallate glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text