Thermo Scientific Chemicals

Chloroform, HPLC Grade, 99.5+% min, Thermo Scientific Chemicals

1 L, Each
Thermo Scientific Chemicals

Chloroform, HPLC Grade, 99.5+% min, Thermo Scientific Chemicals

1 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 022920.K2
also known as 022920-K2
Price (USD)
Price: 83.70
Your Price:
Quantity
-

Chemical Identifiers

CAS
67-66-3
IUPAC Name
trichloromethane
Molecular Formula
CHCl3
InChI Key
HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES
ClC(Cl)Cl
Water Content (Karl Fischer Titration)
≤ 0.03%
Optical Absorbance at
245 nm: ≤ 1.00 AU
Optical Absorbance at
255 nm: ≤ 0.15 AU
Optical Absorbance at
260 nm: ≤ 0.05 AU
Optical Absorbance at
270 nm: ≤ 0.02 AU

Description

Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only