Thermo Scientific Chemicals

Solution tampon pH 11, Thermo Scientific Chemicals Specpure™

Référence: 042424.AP
500 mL, Each
Thermo Scientific Chemicals

Solution tampon pH 11, Thermo Scientific Chemicals Specpure™

Référence: 042424.AP
500 mL, Each
Quantité
Référence: 042424.AP
également connu sous le numéro 042424-AP
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
1336-21-6
Appearance (Color)
Clear colorless
Assay from Supplier's CofA
pH 11.00 (+/- 0.026 at 25°C)
Form
Liquid

Description

Buffer solution (pH 11) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text