Lutetium powder, -40 mesh, 99.9% (REO)
Lutetium powder, -40 mesh, 99.9% (REO)
Lutetium powder, -40 mesh, 99.9% (REO)
Lutetium powder, -40 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Lutetium powder, -40 mesh, 99.9% (REO)

CAS: 7439-94-3 | Lu | 174.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 044167.06
also known as 044167-06
Price (USD)
1,383.65
Special offer
Online exclusive
1,628.00
Save 244.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
1,383.65
Special offer
Online exclusive
1,628.00
Save 244.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7439-94-3
IUPAC Namelutetium
Molecular FormulaLu
InChI KeyOHSVLFRHMCKCQY-UHFFFAOYSA-N
SMILES[Lu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only