Thermo Scientific Chemicals

Tripotassium citrate monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: 045038.36
500 g, Each
Thermo Scientific Chemicals

Tripotassium citrate monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: 045038.36
500 g, Each
Quantity
Catalog number: 045038.36
also known as 045038-36
Price (USD)
Price: 35.50
Online price: 30.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6100-05-6
IUPAC Name
tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Molecular Formula
C6H7K3O8
InChI Key
PJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILES
O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Form
Granular or chunks
Assay from Supplier's CofA
≥96.0%

Description

It is used in the laboratory as a buffering agent and chelator. In renal function studies, potassium citrate is used as an alkanizing agent where the citric acid is a metabolic substrate in renal cells. Studies have shown that potassium citrate may increase bone density, it is therefore used in research experiments related to the treatment of osteoporosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text