This product is now available for online ordering in the U.S. and Canada—Learn more »
Thermo Scientific™
Dionex™ SCC-10 Suppressor Current Controller
Related applications:
Industrial Chromatography
Improve suppressor lifetimes while simultaneously reducing noise in legacy ion chromatography (IC) systems with only four settings for electrolytic suppressor current. The Thermo Scientific™ Dionex™ SCC-10 Suppressor Current Controller provides a selection of 12 current levels so you can choose the optimal operating conditions for your IC applications.
Back to top
The Dionex SCC-10 Suppressor Current Controller easily plugs into legacy system as an external adapter between the suppressor and its power supply. It provides current settings in 12 discrete steps from 10 to 250 mA. Simple to operate, it does not require any changes in the control software.
Compatible Thermo Scientific Legacy Systems
  • Dionex DX-120 Ion Chromatograph
  • Dionex DX-320 and DX-320J Ion Chromatographs
  • Dionex DX-500 Ion Chromatography System
  • Dionex DX-600 Ion Chromatography System
  • Dionex DX-800 Process Analyzer
Note: The Dionex SCC-10 is compatible with the ionex™ SRS™ 300 Self Regenerating Suppressor series; it is not compatible with the Dionex™ CMD™ Carbohydrate Membrane Desalter, SRN™ Self-Regenerating Neutralizer, Atlas™ Suppressor or CES™ Capillary Electrolytic Suppressor series.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.