Interested in a bulk or custom version of this product? Request pricing.

EFNA3 (EphrinA3) Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10188H02H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ephrin-A3 (EFNA3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of human EphrinA3 (NP_004943.1), residues Met 1 to Ser 213, fused to the Fc region of human IgG1 at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized mouseEPHA6 at 1 µg/ml(100 µl/well) can bind human EFNA3-Fc. The ED50 of human EFNA3 is 299.2 ng/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Neurobiology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: EFNA3, EFL2, EPLG3, Ehk1-L, LERK3
Gene ID: 1944
Gene Symbol: EFNA3
Molecular Weight: 48 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Kinases & Inhibitors
Protein Form: Cell Surface Protein, Ligand, Recombinant
Protein Name: ephrin-A3
Protein Subtype: Ephrins
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products