Interested in a bulk or custom version of this product? Request pricing.

F11R (JAMA) Recombinant Human Protein, His Tag

Catalog number: 10198H08H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

F11 Receptor (F11R), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 242) of human F11R (NP_058642.1) fused to the fused Fc region of human IgG1 at the C-terminus.

Activity: Measured by its ability to support the adhesion of Jurkat human leukemic T cells. When 8 x 104 cells/well are added to F11R coated plates (0.2 µg/ml and 100 µl/well), approximately 30-40% will adhere specifically after 60 minutes at 37°C.

Formulation:

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: JAM, FA11R, KAT, JAM1, JAMA, JCAM, CD321, JAM-1, JAM-A, PAM-1
Gene Symbol: F11R
Molecular Weight: 25 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Other Proteins
Protein Name: F11 receptor
Protein Subtype: Intercellular adhesion Molecule, Viral Host Cell Receptors
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: Other Research
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols