Interested in a bulk or custom version of this product? Request pricing.

FCGR3A (CD16a), 176 Phe/Val Recombinant Human Protein, His Tag

Catalog number: 10389H08H150

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Fc gamma receptor III-a (FCGR3A/CD16a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 208) of human FCGR3A (P08637-1), containing the mutation 176 Phe/Val, precursor fused to a polyhistidine tag at the C-terminus.

Activity: Measured using the Octet RED System, the affinity constant (Kd) of FCGR3A bound to human IgG1 was 180nM.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins, Growth Factors & Receptors
Research Category: Immunology, Inflammation, Neurobiology, Virology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: RP11-5K23.1, CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Gene ID: 2214
Gene Symbol: FCGR3A
Molecular Weight: 23.3 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: Fc fragment of IgG, low affinity IIIa, receptor (CD16a)
Protein Subtype: Fc Receptors, Glycoproteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols