Interested in a bulk or custom version of this product? Request pricing.

EGF Recombinant Human Protein

Catalog number: 10605HNAE250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Manufactured by Sino Biological (China); Distributed by Life Technologies.

Epidermal growth factor (EGF) recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of human EGF (NP_001954.2) (Asn 971-Arg 1023) retaining the initial Met at the N-terminus.

N-terminal Sequence Analysis: Met 1

Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblasts (Rubin, J.S. et al. 1991, Proc. Natl. Acad. Sci. USA 88:415.) . The EC50 for this effect is typically 20-100 pg/mL.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween®-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

Background
Human epidermal growth factor (EGF) exerts actions on a wide variety of biological processes, and plays an important role in the regulation of cell growth, proliferation, and differentiation mediated by its high-affinity receptor EGFR on the cell surface. It induces differentiation of specific cells, promotes angiogenesis, and inhibits gastric acid secretion in vivo, while serves as a potent mitogenic actor for a variety of cultured cells of both ectodermal and mesodermal origin. Human EGF was originally isolated from urine based on its inhibitory effect on gastric secretion and named urogastrone accordingly. EGF is the founding member of the EGF-family of proteins which are characterized by the presence of at least one EGF structural unit (a conserved 6 cysteine motif that forms three disulfide bonds). The EGF precursor is believed to exist as a membrane-bound molecule containing 9 EGF units, and is proteolyticallycleaved to generate the 53-amino acid peptide hormone.

You may also be interested in the following EGF products:
Kinases
Primary Antibodies
Bioconjugates
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Angiogenesis, Cell Cycle & Proliferation
Expression: E. coli
Form: Lyophilized
Gene Aliases: EGF, HOMG4, URG
Gene Symbol: EGF
Molecular Weight: 6.3 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Growth Factors & Receptors
Protein Form: Full Length, Recombinant
Protein Name: Epidermal growth factor
Protein Subtype: EGF (Epidermal Growth Factor)
Protein Tag or Fusion: Untagged
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products