Interested in a bulk or custom version of this product? Request pricing.

IGF2BP2 Recombinant Human Protein, N-His.GST Tag

Catalog number: 11116H20B5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Insulin-like Growth Factor 2 mRNA Binding Protein 2 (IGF2BP2), recombinant human protein is supplied as a lyophilized powder.

This recombinant protein was expressed from a DNA sequence encoding the human IGF2BP2 isoform 1 (Q9Y6M1-2) (Met 1-Lys 599) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 50 mM Tris, 500 mM NaCl , 0.5 mM PMSF, 5 mM GSH, pH8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Apoptosis, Development, Neurobiology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Lyophilized
Gene Aliases: IMP-2, IMP2, VICKZ2, p62
Gene ID: 10644
Gene Symbol: IGF2BP2
Molecular Weight: 94 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Family: Growth Factors & Receptors
Protein Form: Full Length, Recombinant
Protein Name: Insulin-like Growth Factor 2 mRNA Binding Protein 2
Protein Tag or Fusion: GST Tag, His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Tag Position: N-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products