Product Image
Gibco™

HuMEC Basal Serum-Free Medium (1X)

HuMEC Basal Serum-Free Medium is designed to work with HuMEC Supplement Kit (SKU# 12755013). Together, these products are engineered specificallyRead more
Have Questions?
Catalog NumberQuantity
12753018500 mL
Catalog number 12753018
Price (USD)
163.00
Each
Quantity:
500 mL
Price (USD)
163.00
Each
HuMEC Basal Serum-Free Medium is designed to work with HuMEC Supplement Kit (SKU# 12755013). Together, these products are engineered specifically for the culture of human mammary epithelial cells (SKU# A10565). This translates to greater experimental validity, more reproducible results, and reduction of lot-to-lot variability and aberrations that can occur with the addition of serum. Human mammary epithelial cells cultured in HuMEC Medium have gene expression profiles consistent with normal mammary epithelial cells, as tested on a cDNA microarray.

Applications:
HuMEC Medium is optimized specifically for the growth of human mammary epithelial cells, and has been performance tested in a growth assay using normal human mammary epithelial cells. HuMEC Medium promotes superior growth of normal human mammary epithelial cells, making it ideal for researchers in breast cancer, drug discovery, and other aspects of breast biology.

Contents and Storage:
HuMEC Basal Serum Free Medium should be supplemented with HuMEC Supplement Kit. HuMEC Supplement Kit and HuMEC Basal Serum Free Media can be ordered together as HuMEC Ready Medium (SKU# 12752010). The HuMEC Supplement Kit includes 5 ml of a supplement mix containing epidermal growth factor, hydrocortisone, isoproterenol, transferrin, and insulin, and 25 mg of bovine pituitary extract. Store the basal medium at +2°C to +8°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeEpithelial Cell (Mammary)
Concentration1 X
Product TypeHuMEC Basal Serum-Free Medium
Quantity500 mL
Shelf Life16 Months From Date of Manufacture
ClassificationSerum-free
FormLiquid
SterilitySterile-filtered
With AdditivesPhenol Red
Unit SizeEach