Thermo Scientific™
Peristaltic Pump Tubing
Related applications:
Industrial Mass Spectrometry
Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ Q ICP-MS; all designed for durability and fitness for purpose.
Back to top
Select High Quality Consumables for Improved Performance
Choose from the following Peristaltic Pump Tubing:
  • Peristaltic Pump Tubing - dia.0,508 -Orange/Yellow -PVC
  • Peristaltic Pump Tubing - dia.0,381 -Orange/Green - PVC
  • Peristaltic Pump Tubing - dia.0,762 - Black/Black -PVC
  • Peristaltic Pump Tubing - dia.0,254 - Orange/Blue -PVC
  • Peristaltic Pump Tubing - dia.1,295 - Gray/Gray -Santoprene
  • Peristaltic Pump Tubing - dia.3,175 - Black/White -Santoprene