Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
1343390 | Speciation Kit for ICS-2100 system |
1343380 | Speciation Kit for ICS-900 system |
1343400 | Speciation Kit for ICS-5000+ system |
Advance your laboratorys capabilities with Thermo Scientific™ IC-ICP-MS Speciation Kits. Use with the Thermo Scientific™ iCAP™ Q ICP-MS system to enable routine elemental speciation analysis.
With all the necessary parts for simplified speciation, these kits also include dedicated software plug-ins in the Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ software for iCAP Q ICP-MS systems, giving complete bi-directional control of coupled systems without the need for an extra software package or hardware triggering.
Available Kits: