Pax5 Mouse Monoclonal Antibody (clone ZP007)

Catalog number:  180472

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This monoclonal antibody reacts with Pax5. Immunogen source: Recombinant human Pax5. Positive tissue: Tonsil, B cell lymphoma

Clone_PAD
ZP007

Immunogen
Immunogen source: Recombinant human Pax5.

Specificity
Pax5

Reactivity
Human

Applications
Immunohistochemistry (FFPE)

Pretreatment
Epitope Retrieval: Required (EDTA pH 8.0)

Staining Pattern
Nuclear

Storage
2-8°C

RUO_IVD
IVD

CE
For In Vitro Diagnostic Use.

Specifications

SubClass (Isotype): IgG1, kappa
Clonality: Monoclonal
Label or Dye: Unconjugated
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols