1,1'-Azobis(N,N-dimethylformamide), 98%, Thermo Scientific Chemicals
1,1'-Azobis(N,N-dimethylformamide), 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1'-Azobis(N,N-dimethylformamide), 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 185710050
Price (USD)/ Each
405.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
477.00 
Save 71.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
405.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
477.00 
Save 71.35 (15%)
Add to cart
1,1'-Azobis(N,N-dimethylformamide), 98%, Thermo Scientific Chemicals
Catalog number185710050
Price (USD)/ Each
405.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
477.00 
Save 71.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-[(dimethylcarbamoyl)imino]-3,3-dimethylurea
Molecular FormulaC6H12N4O2
InChI KeyVLSDXINSOMDCBK-UHFFFAOYSA-N
SMILESCN(C)C(=O)N=NC(=O)N(C)C
Molecular Weight (g/mol)172.19
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point104°C to 114°C
GC>=97.5 %
Appearance (Color)Yellow
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only