CaptureSelect™ LC-kappa (mur) Affinity Matrix
Thermo Scientific™

CaptureSelect™ LC-kappa (mur) Affinity Matrix

Thermo Scientific™ CaptureSelect™ LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
19131501010 mL
1913150055 mL
19131505050 mL
Catalog number 191315010
Price (USD)
2,300.00
Each
Quantity:
10 mL
Price (USD)
2,300.00
Each

CaptureSelect LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins. The ligand is directed towards a unique domain on the constant part of the kappa light chain of immunoglobulins, which makes CaptureSelect LC-kappa (mur) Affinity Matrix a unique resin in terms of specificity.

This matrix can be used for the purification and isolation of kappa light chain-containing murine immunoglobulins and Fab fragments from complex sources like plasma, serum, and cell culture supernatants in a single step. Due to its non-reactivity with bovine Ig, FCS can be applied in growth media without disturbing purification.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 mL
Stationary PhaseAnti-LC-kappa (mur)
Capacity (Metric)>15 g/L resin
FormResin Slurry
Particle Size35 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light