Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl, Thermo Scientific Chemicals
 
Have Questions?
Quantity:
500 mL
Packaging:
Plastic bottle
Catalog number 196465000
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
118.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
118.00 
Save 17.35 (15%)
Add to cart
Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl, Thermo Scientific Chemicals
Catalog number196465000
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
118.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-66-6, 7647-01-0, 7732-18-5, 7646-85-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength990 to 1010 µg/ml of Zn(2+) in 2-5% HCl
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only