Reagents and Bottle Kits for Orion™ 2030 Silica Analyzers
Thermo Scientific™
Reagents and Bottle Kits for Orion™ 2030 Silica Analyzers
Avoid productivity lapses by ensuring your Thermo Scientific™ Orion™ 2030 Silica Analyzer has the reagents and standards it needs.
Have Questions?
Change view
Catalog Number
Description
203020
Silica 200 ppb Standard, 5L
203002
Silica 20 ppb Standard, 4 x 475mL
203030
Silica 1 ppm Standard, 5L
Catalog number 203020
Price (USD)/ Each
-
Request A Quote
Description:
Silica 200 ppb Standard, 5L
Avoid productivity lapses by ensuring your Thermo Scientific™ Orion™ 2030 Silica Analyzer has the reagents and standards it needs. Thermo Scientific™ Reagents and Bottle Kits are used with the Orion™ 2030 Silica Analyzer.
Includes:
Silica kit available with or without reagents.
With reagents: 5L each of reagents 1, 2 and 3 with caps and tubing
Without reagents: 3 x 5L empty bottles for reagents 1, 2 and 3; reagent bottle caps and reagent tubing