Orion™ 2110XP Ammonia Analyzer
Actual product may vary
Thermo Scientific™

Orion™ 2110XP Ammonia Analyzer

Control and optimize boiler ammonia levels with continuous, direct and precise ammonia measurements from the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2110XPAmmonia Analyzer
2110XPENAnalyzer with Enclosure
Catalog number 2110XP
Price (MXN)
-
Type:
Ammonia Analyzer
Reduce operational costs while producing the highest purity steam with the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer. This analyzer provides continuous, direct and precise measurements for the control and optimization of boiler ammonia levels.

Features

  • Accurate detection allows precise ammonia control with confidence — maintain proper chemical feed levels without the guess work or excessive operating cost.
  • Stable and drift-free measurements with minimal maintenance eliminate the need for frequent calibrations, saving you time and money.
  • Repeatable and verifiable ammonia measurements using ammonia electrode technology for the highest level of performance available.
  • Designed for ammonia specificity, the unique chemistry provides unsurpassed results across a wide range without the pitfalls of pH and conductivity 'bottom out effects.'
  • Step-by-step calibration process ensures a successful calibration the first time and every time.
  • Ammonia detection range of 0 to 10ppm provides direct, accurate and precise results.
  • Continuous online measurement ensures real-time information for optimum control of the ammonia feed without excessive operating costs.
  • Selective and sensitive for ammonia, our system surpasses the performance of other systems that have the limitations of pH and conductivity to track real-time boiler ammonia levels.

2110XP Ammonia Analyzer Ordering Information

  • Annual Consumables Kit, Cat. No. 211050XP
  • Reagent, Cat. No. 181030
  • Diffusion Tubing, Cat. No. 150060
  • Calibration Kit, Cat. No. 2110XPCAL
  • Calibration Standards, Cat. No. 181040
  • Ammonia Electrode, Cat. No. 100047
  • Reference Electrode, Cat. No. 210056
  • Reference Electrode Filling Solution, Cat. No. 181073
Specifications
Accuracy±15% or 30 ppb, whichever is greater
Accuracy (Temperature)±0.5°C
Accuracy (mV)± (0.5 mV + 0.1%)
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
CalibrationDouble Known Addition
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
Channel 1Ammonia
Channel 2Optional ph/ORP or Conductivity
Channels1
Communication DetailsModbus with Additional Module
Current Loops2
Description2110XP Ammonia Analyzer
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
Includes2110XP ammonia analyzer with Ammonia electrode, reference electrode (100056), ATC probe (2100TP),ammonia calibration standards kit, CD user guide and options kit
Inlet Pressure8 to 100psig
Inputs85-132; 170-264 VAC
Keypad TypeMembrane
Length (English)11 in.
Length (Metric)27 cm
Max. Relative Humidity5 to 95%, non-condensing
Measurement Ranges0 to 10ppm
Mounting OptionsWall mount
Outputs0 to 20 mA; 4 to 20 mA
Range (mV)±1999.9m
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Resolution (Temperature)0.1°C
Resolution (mV)0.1mV
Response Time95% within 2 minutes of injecting a standard solution
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Sample Temperature5 to 45°C
Voltage85/132 VAC, 170/264 VAC
Width (English)17 in.
Width (Metric)45 cm
Temperature (Metric) Operating5°C to 35°C
TypeAmmonia Analyzer
Unit SizeEach