EZ-Link™ Iodoacetyl-LC-Biotin
Thermo Scientific™

EZ-Link™ Iodoacetyl-LC-Biotin

Thermo Scientific EZ-Link Iodoacetyl-LC-Biotin is a mid-length, haloacetyl-activated, sulfhydryl-reactive biotinylation reagent that forms stable, irreversible thioether bonds at alkaline pH.FeaturesRead more
Have Questions?
Catalog NumberQuantity
2133350 mg
Catalog number 21333
Price (USD)
458.00
Each
Quantity:
50 mg
Price (USD)
458.00
Each
Thermo Scientific EZ-Link Iodoacetyl-LC-Biotin is a mid-length, haloacetyl-activated, sulfhydryl-reactive biotinylation reagent that forms stable, irreversible thioether bonds at alkaline pH.

Features of EZ-Link Iodoacetyl-LC-Biotin:

• Protein labeling—biotinylate antibodies or other proteins for use in protein methods
• Membrane-permeable—can be used to label inside cells (intracellular)
• Thiol-reactive—reacts with sulfhydryls (-SH), such as the side-chain of cysteine (C)
• Iodoacetyl-activated—perform reactions in the dark at pH 7.5 to 8.5 in Tris or borate buffer
• Irreversible—forms permanent thioether bonds; spacer arm cannot be cleaved
• Solubility—must be dissolved in DMSO or DMF before further dilution in aqueous buffers
• Medium length—spacer arm (total length added to target) is 27.1 angstroms; contains hexylenediamine extension

Iodoacetyl-LC-Biotin is a haloacetyl-biotin compound for labeling protein cysteines and other molecules that contain sulfhydryl groups. This reagent specifically reacts with reduced thiols (-SH) in alkaline buffers to form permanent (irreversible) thioether bonds. The unique feature of Iodoacetyl-LC-Biotin is its extended yet chemically simple hexylenediamine spacer arm.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.

Biotinylation reagents differ in reactivity, length, solubility, cell permeability and cleavability. Three types of sulfhydryl-reactive compounds are available: maleimido, iodoacetyl and pyridyldithiol. Iodoacetyl reagents specifically react with sulfhydryl groups (-SH) at pH 8.3 to form permanent thioether bonds.

In proteins, sulfhydryls exist where there are cysteine (C) residues. Cystine disulfide bonds must be reduced to make sulfhydryl groups available for labeling. Hinge-region disulfide bridges of antibodies can be selectively reduced to make functional half-antibodies that can be labeled.

Applications:
• Electron microscopy studies on spatial relationships between proteins (Ref.1)
• Localizing the SH1 thiol of the myosin head using avidin-biotin complexes in electron microscopy (Ref.2)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-Permeant
Label TypeBiotin & Analogs
Product LineEZ-Link
Product TypeIodoacetyl-LC-Biotin
Quantity50 mg
Reactive MoietyHaloacetyl
Chemical ReactivityThiol
Label or DyeBiotin
SolubilityDMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
SpacerLong
Unit SizeEach
Contents & Storage
Store desiccated at 4°C. Shipped at ambient temperature.

Frequently asked questions (FAQs)

What is the advantage of using EZ-Link HPDP-Biotin over maleimide- or iodoacetyl-containing biotinylation reagents?

Both maleimide- and iodoacetyl-containing biotinylation reagents react with sulfhydryl (-SH) groups to form stable thioether bonds that are not cleavable. EZ-Link HPDP-Biotin on the other hand is a pyridyldithiol-biotin compound that reacts with -SH groups in near-neutral buffers to form reversible disulfide bonds. Because the disulfide group can be cleaved using DTT or other reducing agents, HPDP-Biotin is useful for labeling and affinity-purification applications that require recovery of the original, unmodified molecule.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (1)

Citations & References
Abstract
Dysregulated Redox Regulation Contributes to Nuclear EGFR Localization and Pathogenicity in Lung Cancer.
Authors:Little AC, Hristova M, van Lith L, Schiffers C, Dustin CM, Habibovic A, Danyal K, Heppner DE, Lin MJ, van der Velden J, Janssen-Heininger YM, van der Vliet A
Journal:Sci Rep
PubMed ID:30890751
Lung cancers are frequently characterized by inappropriate activation of epidermal growth factor receptor (EGFR)-dependent signaling and epigenetic silencing of the NADPH oxidase (NOX) enzyme DUOX1, both potentially contributing to worse prognosis. Based on previous findings linking DUOX1 with redox-dependent EGFR activation, the present studies were designed to evaluate whether DUOX1 ... More