KingFisher™ Flex 24 Deep Well head
Thermo Scientific™

KingFisher™ Flex 24 Deep Well head

This is a magnetic head for the KingFisher™ Flex Magnetic Particle Purification System (Cat. Nos. 5400630 and 5400640). This magneticRead more
Have Questions?
Catalog NumberQuantity
24074440Each
Catalog number 24074440
Price (USD)
5,367.65
Online exclusive
6,170.00
Save 802.35
Each
Quantity:
Each
Price (USD)
5,367.65
Online exclusive
6,170.00
Save 802.35
Each
This is a magnetic head for the KingFisher™ Flex Magnetic Particle Purification System (Cat. Nos. 5400630 and 5400640). This magnetic head can serve as a replacement head for the 24 deep-well format system or a supplemental head for the 96 deep-well format system.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)KingFisher™ Flex
Product LineKingFisher
QuantityEach
Shipping ConditionRoom Temperature
Capacity24 Samples
Format24 Deep-well Head
Unit SizeEach

Frequently asked questions (FAQs)

I see that the KingFisher 24 Deep Well Head has a processing volume of 5 mL. Does that mean that I can process up to 5 mL of my sample?

No, the processing volume refers to the maximum volume that can be held in the 24 Deep Well plates. This incorporates the volume of binding buffers and beads, so when scaling up reaction volumes, it is important to make sure that the total volume does not exceed 5 mL.

My KingFisher Flex run was stopped due to an error message. Can I start the protocol from where it stopped?

No, once a protocol was stopped, it will restart from the beginning. The extraction procedure can either be finished manually by using a magnetic stand, or a modified version of the original KingFisher Flex script would have to be created that starts off at the point where the run was stopped.

I have a 12-plate protocol but there are only 8 slots on the KingFisher Flex instrument turntable. What should I do with the remaining 4 plates?

Set up the protocol on the BindIt software. After the first 8 plates are done, the instrument will pause and instruct you to switch out the plates.

Is it always necessary to use the transport locks on the KingFisher Flex instrument?

The transport locks are only necessary when relocating the instrument.

Are the volumes of reagents in each well critical for a successful KingFisher Flex run?

We strongly recommend that you maintain the specified volumes within the defined limits to avoid spillover and to help ensure the best performance at the most efficient level.