Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Red |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Height (Metric) | 224 mm |
Product Type | Color-coded Wash Bottle |
Height (English) | 8.8 in. |
Closure Material | Polypropylene |
Closure Size | 28 mm |
Diameter (Metric) | 72.9 mm |
Closure Type | Screw Cap |
Through the Closure Dispensing | Yes |
Unit Size | Pack of 6 |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Yellow |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Height (Metric) | 224 mm |
Product Type | Color-coded Wash Bottle |
Height (English) | 8.8 in. |
Closure Material | Polypropylene |
Closure Size | 28 mm |
Diameter (Metric) | 72.9 mm |
Closure Type | Screw Cap |
Through the Closure Dispensing | Yes |
Unit Size | Case of 24 |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Yellow |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Height (Metric) | 224 mm |
Product Type | Color-coded Wash Bottle |
Height (English) | 8.8 in. |
Closure Material | Polypropylene |
Closure Size | 28 mm |
Diameter (Metric) | 72.9 mm |
Closure Type | Screw Cap |
Through the Closure Dispensing | Yes |
Unit Size | Pack of 6 |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Blue |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Height (Metric) | 224 mm |
Product Type | Color-coded Wash Bottle |
Height (English) | 8.8 in. |
Closure Material | Polypropylene |
Closure Size | 28 mm |
Diameter (Metric) | 72.9 mm |
Closure Type | Screw Cap |
Through the Closure Dispensing | Yes |
Unit Size | Case of 24 |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Blue |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Height (Metric) | 224 mm |
Product Type | Color-coded Wash Bottle |
Height (English) | 8.8 in. |
Closure Material | Polypropylene |
Closure Size | 28 mm |
Diameter (Metric) | 72.9 mm |
Closure Type | Screw Cap |
Through the Closure Dispensing | Yes |
Unit Size | Pack of 6 |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Red |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Height (Metric) | 224 mm |
Product Type | Color-coded Wash Bottle |
Height (English) | 8.8 in. |
Closure Material | Polypropylene |
Closure Size | 28 mm |
Diameter (Metric) | 72.9 mm |
Closure Type | Screw Cap |
Through the Closure Dispensing | Yes |
Unit Size | Case of 24 |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Green |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Height (Metric) | 224 mm |
Product Type | Color-coded Wash Bottle |
Height (English) | 8.8 in. |
Closure Material | Polypropylene |
Closure Size | 28 mm |
Diameter (Metric) | 72.9 mm |
Closure Type | Screw Cap |
Through the Closure Dispensing | Yes |
Unit Size | Case of 24 |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
Description | With stem and draw tube |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 6 |
Closure Color | Green |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Mouth | Narrow |
Shape | Cylindrical |
Height (Metric) | 224 mm |
Product Type | Color-coded Wash Bottle |
Height (English) | 8.8 in. |
Closure Material | Polypropylene |
Closure Size | 28 mm |
Diameter (Metric) | 72.9 mm |
Closure Type | Screw Cap |
Through the Closure Dispensing | Yes |
Unit Size | Pack of 6 |
Catalog Number | Specifications | Unit Size | Closure Color | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2422-5500PK | Pack of 6 | Red | 71.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2422-2500 | Case of 24 | Yellow | 242.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2422-2500PK also known as 2422-2500 | Pack of 6 | Yellow | 71.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2422-3500 | Case of 24 | Blue | 242.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2422-3500PK also known as 2422-3500 | Pack of 6 | Blue | 71.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2422-5500 | Case of 24 | Red | 242.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2422-4500 | Case of 24 | Green | 242.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2422-4500PK also known as 2422-4500 | Pack of 6 | Green | 71.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|