Dionex™ Guardcap™ Na Vial Caps
Actual product may vary
Dionex™ Guardcap™ Na Vial Caps
Dionex™ Guardcap™ Na Vial Caps
Dionex™ Guardcap™ Na Vial Caps
Dionex™ Guardcap™ Na Vial Caps
Thermo Scientific™
Dionex™ Guardcap™ Na Vial Caps
Remove cations and metals from IC samples without additional labor, time, and complexity using Thermo Scientific™ Dionex™ Guardcap™ Na vial caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
302797Functionalized Vial Cap
302796Functionalized Vial Cap with Transfer Line
Catalog number 302797
Price (USD)
479.00
Each 1
Product Type:
Functionalized Vial Cap
Price (USD)
479.00
Each 1
Remove high levels of alkaline earth and transition metals without changing sample pH and filter particulates by using Thermo Scientific™ Dionex™ Guardcap™ Na vial caps. Designed for use with Thermo Scientific™ Dionex™ AS-DV autosampler PolyVials, Dionex Guardcap Na vial caps contain sulfonated strong cation exchange resin in the sodium form embedded in a high density polyethylene matrix. Dionex Guardcap Na vial caps seamlessly bring the power of automated inline sample preparation to your existing sample workflow without the need for additional labor, time, and complexity.
Each Guardcap Na vial cap:
  • Removes 1000mg/L divalent cation (e.g., calcium or transition metals) from 800μL of sample.
  • Leaves sample pH unchanged.
  • Filters better than 80% of particulate matter greater than 20μm in diameter.
Ideal for determination of pH-sensitive analytes
Some analytes, most notably nitrite, are affected by the pH change caused by hydronium-form resin. Removal of metals from a sample requiring determination of nitrite must be accomplished through sodium-form resin to ensure quantitative recovery (> 99%).
Obtain superior analytical results with greatly reduced effort
Increase the lifetime of your analytical columns and suppressors by removing metals, matrix ions, and particulates that can foul them.
Convenient and easy to use
Dionex Guardcap Na vial caps come in the familiar filter cap format used with Dionex PolyVials. Unlike traditional offline sample preparation cartridges, Dionex Guardcap Na vial caps require no time-consuming pretreatment procedures prior to use ̶ simply install these functionalized vial caps on your sample vials and your samples are ready for analysis.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionDionex Guardcap Na Vial Caps
For Use With (Equipment)5 mL AS-DV PolyVials
MaterialOnGuard Na Resin
QuantityEach 1
Product TypeFunctionalized Vial Cap
Unit SizeEach 1