Regulatory requirements vary by country; products may not be available in your geographic area.
PrioSCAN® Package
Catalog number: 30900
 Related applications: Animal Health
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The PrioSCAN® is a one-button solution for the fast read out and interpretation of PrioSTRIP™ and PrioSTRIP™ SR results. The results are stored as a PDF-file and can be exported to an Excel spreadsheet or a laboratory information management system. An entire 96-plate can be analyzed in one run. PrioSCAN® detects the blue lines on the strips of the PrioSTRIP™ combs and translates the information into digital data. The data points are evaluated and interpreted with the PrioSCAN® analysis software. This package product consists of a scanner and laptop.
For Veterinary Use Only.
Specifications
For Use With (Equipment): 30000 PrioSTRIP™
Product Line: PrioSCAN®, Prionics™
Product Size: 1 piece
Documents
Manuals & protocols