Thermo Scientific™

Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap

Catalog number: 3139-0016PK
Thermo Scientific™

Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Cap

Catalog number: 3139-0016PK
Spin with confidence with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes with Sealing Caps. These tubes are chemically compatible with a wider variety of lab reagents than polycarbonate centrifuge tubes and are repeately autoclavable for extended performance.
 
Catalog Number
3139-0016PK
also known as 3139-0016
Unit Size
Pack of 10
Diameter (Metric)
18.1 mm
Capacity (Metric)
16 mL
Length (Metric)
106.4 mm
Max. RCF
50,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene, red rubber
Capacity (Metric)16 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)106.4 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)18.1 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizePack of 10
Catalog Number
Unit Size
Case of 50
Diameter (Metric)
16.1 mm
Capacity (Metric)
10 mL
Length (Metric)
82.8 mm
Max. RCF
50,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene, red rubber
Capacity (Metric)10 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)82.8 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)16.1 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizeCase of 50
Catalog Number
3139-0010PK
also known as 3139-0010
Unit Size
Pack of 10
Diameter (Metric)
16.1 mm
Capacity (Metric)
10 mL
Length (Metric)
82.8 mm
Max. RCF
50,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene, red rubber
Capacity (Metric)10 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)82.8 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)16.1 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizePack of 10
Catalog Number
Unit Size
Case of 50
Diameter (Metric)
18.1 mm
Capacity (Metric)
16 mL
Length (Metric)
106.4 mm
Max. RCF
50,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene, red rubber
Capacity (Metric)16 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)106.4 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)18.1 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizeCase of 50
Catalog Number
Unit Size
Case of 50
Diameter (Metric)
25.8 mm
Capacity (Metric)
30 mL
Length (Metric)
102.1 mm
Max. RCF
50,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene, red rubber
Capacity (Metric)30 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)102.1 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)25.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizeCase of 50
Catalog Number
3139-0030PK
also known as 3139-0030
Unit Size
Pack of 10
Diameter (Metric)
25.8 mm
Capacity (Metric)
30 mL
Length (Metric)
102.1 mm
Max. RCF
50,000 x g
Price (USD)
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene, red rubber
Capacity (Metric)30 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)102.1 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)25.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizePack of 10
Catalog Number
Unit Size
Case of 50
Diameter (Metric)
28.8 mm
Capacity (Metric)
50 mL
Length (Metric)
113.5 mm
Max. RCF
100,605 x g in the T29-8x50 rotor
Price (USD)
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene, red rubber
Capacity (Metric)50 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)113.5 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)28.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF100,605 x g in the T29-8x50 rotor
Unit SizeCase of 50
Catalog Number
3139-0050PK
also known as 3139-0050
Unit Size
Pack of 10
Diameter (Metric)
28.8 mm
Capacity (Metric)
50 mL
Length (Metric)
113.5 mm
Max. RCF
100,605 x g in the T29-8x50 rotor
Price (USD)
Availability
-
Full specifications
Quantity
Closure MaterialPolypropylene, red rubber
Capacity (Metric)50 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)113.5 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)28.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF100,605 x g in the T29-8x50 rotor
Unit SizePack of 10
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeDiameter (Metric)Capacity (Metric)Length (Metric)Max. RCFPrice (USD)AvailabilityQuantity
3139-0016PK
also known as 3139-0016
Full specifications
Pack of 1018.1 mm16 mL106.4 mm50,000 x g
-
Closure MaterialPolypropylene, red rubber
Capacity (Metric)16 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)106.4 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)18.1 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizePack of 10
3139-0010Full specifications
Case of 5016.1 mm10 mL82.8 mm50,000 x g
-
Closure MaterialPolypropylene, red rubber
Capacity (Metric)10 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)82.8 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)16.1 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizeCase of 50
3139-0010PK
also known as 3139-0010
Full specifications
Pack of 1016.1 mm10 mL82.8 mm50,000 x g
-
Closure MaterialPolypropylene, red rubber
Capacity (Metric)10 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)82.8 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)16.1 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizePack of 10
3139-0016Full specifications
Case of 5018.1 mm16 mL106.4 mm50,000 x g
-
Closure MaterialPolypropylene, red rubber
Capacity (Metric)16 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)106.4 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)18.1 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizeCase of 50
3139-0030Full specifications
Case of 5025.8 mm30 mL102.1 mm50,000 x g
-
Closure MaterialPolypropylene, red rubber
Capacity (Metric)30 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)102.1 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)25.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizeCase of 50
3139-0030PK
also known as 3139-0030
Full specifications
Pack of 1025.8 mm30 mL102.1 mm50,000 x g
-
Closure MaterialPolypropylene, red rubber
Capacity (Metric)30 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)102.1 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)25.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g
Unit SizePack of 10
3139-0050Full specifications
Case of 5028.8 mm50 mL113.5 mm100,605 x g in the T29-8x50 rotor
-
Closure MaterialPolypropylene, red rubber
Capacity (Metric)50 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)113.5 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)28.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF100,605 x g in the T29-8x50 rotor
Unit SizeCase of 50
3139-0050PK
also known as 3139-0050
Full specifications
Pack of 1028.8 mm50 mL113.5 mm100,605 x g in the T29-8x50 rotor
-
Closure MaterialPolypropylene, red rubber
Capacity (Metric)50 mL
MaterialPolypropylene Copolymer
No. per Case50
No. per Pack10
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Length (Metric)113.5 mm
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)28.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF100,605 x g in the T29-8x50 rotor
Unit SizePack of 10
Showing 8 of 8
Features
  • Greater chemical resistance than polycarbonate
  • Can be autoclaved repeatedly—last longer than polycarbonate tubes under conditions of repeated autoclaving
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges; 3139-0050 can be used up to 100,605 x g in the T29-8x50 rotor
  • Come with polypropylene sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Must be filled to 80% of total capacity when centrifuging
  • Autoclavable/Leakproof† when used with supplied Nalgene closure; Repeated autoclaving will reduce the life of PC centrifuge tubes. 

Ordering Information:

Replacement sealing-cap assemblies available for selected sizes. Use DS3131-0020 for 30mL tubes and DS3131-0024 for 50mL tubes. Actual brim capacity of 3139-0050 is 42mL but is designed to fit standard rotor.



Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text