Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Color | Description | Sterility | Fit for Purpose | Unit Size |
---|---|---|---|---|---|
3470NKS | Natural | 2.0ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3422NK | Natural | 0.5ml Screw Cap Tube, NonKnurl,Skirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3472NK | Natural | 0.5ml Screw Cap Tube, NonKnurl,Skirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3465NK | Natural | 0.5ml Screw Cap Tube, NonKnurl,Skirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3422NKS | Natural | 0.5ml Screw Cap Tube, NonKnurl,Skirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3462NK | Natural | 0.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3460NK | Natural | 0.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3431NK | Natural | 0.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3462NKS | Natural | 0.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3422-11 | Natural | 0.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3422-11PK also known as 3422-11 | Natural | 0.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Pack of 500 |
3472 | Natural | 0.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3465 | Natural | 0.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3465PK | Natural | 0.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Pack of 500 |
3422S | Natural | 0.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3466NK | Natural | 1.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3464NK | Natural | 1.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3461NK | Natural | 1.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3466NKS | Natural | 1.5ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3478 | Natural | 1.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3478PK also known as 3478 | Natural | 1.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Pack of 500 |
3474 | Natural | 1.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3474PK also known as 3474 | Natural | 1.5ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Pack of 500 |
3467-11 | Natural | 1.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3478S | Natural | 1.5ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3466 | Natural | 1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3466PK also known as 3466 | Natural | 1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Pack of 500 |
3464 | Natural | 1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3464PK also known as 3464 | Natural | 1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Pack of 500 |
3461 | Natural | 1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3466S | Natural | 1.5ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3490NK | Natural | 2.0ml Screw Cap Tube, NonKnurl,Skirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3488NK | Natural | 2.0ml Screw Cap Tube, NonKnurl,Skirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3469NK | Natural | 2.0ml Screw Cap Tube, NonKnurl,Skirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3490NKS | Natural | 2.0ml Screw Cap Tube, NonKnurl,Skirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3470NK | Natural | 2.0ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3468NK | Natural | 2.0ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3463NK | Natural | 2.0ml Screw Cap Tube, NonKnurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3490 | Natural | 2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3490PK also known as 3490 | Natural | 2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Pack of 500 |
3488 | Natural | 2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3488PK also known as 3488 | Natural | 2.0ml Screw Cap Tube,Knurl,Skirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Pack of 500 |
3469-11 | Natural | 2.0ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3469-11PK also known as 3469-11 | Natural | 2.0ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Pack of 500 |
3490S | Natural | 2.0ml Screw Cap Tube,Knurl,Skirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3470 | Natural | 2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Case of 5000 |
3470PK | Natural | 2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube only no cap | Non-sterile | PCR Ready | Pack of 500 |
3468 | Natural | 2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Case of 5000 |
3468PK also known as 3468 | Natural | 2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, Non-Sterile tube w/ unattached cap | Non-sterile | PCR Ready | Pack of 500 |
3463 | Natural | 2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3463PK also known as 3463 | Natural | 2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile tube w/ attached cap | Sterile | Sterile & PCR Ready | Pack of 500 |
3470S | Natural | 2.0ml Screw Cap Tube,Knurl, NonSkirted,Natural, E-Beam Sterile, tube only no cap | Sterile | Sterile & PCR Ready | Case of 5000 |
3467TSPK also known as 3467 | Natural | Free-standing tube with screw cap & o-ring | Sterile | Sterile & PCR Ready | Pack of 500 |
Choose Thermo Scientific™ Screw Cap Micro Tubes, which available in conical and free-standing designs, when seeking secure, short-term sample storage in low temperatures. Their robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear, externally threaded tubes and UV protective amber tubes. Options include: with or without a self-standing skirted bottom, with or without tethered straps, non-knurl and knurled tubes for single-hand operation, choice of multicolored caps and volume sizes of 0.5mL, 1.5mL and 2.0mL sizes. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.
Non-Sterile is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor.
Sterile E-Beam is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, ATP and PCR Inhibitor.
Packaging is tailored to meet broad requirements. Tubes are packaged with and without caps, caps are attached or unattached, placed in a single bag and double bagged feature for clean room environments is available upon request.
100% virgin polypropylene resin specifically selected for high clarity and biologic applications.
O-ring design, thickness and material selection have been optimized to provide protective features for maximum sealing ability, biologic interaction and temperature capability. The black colored O-ring is made from EPDM.
Automated System Compatible. Whether automate is used for filling, capping or labeling, the product is designed to withstand high torque pressures associated with automation. Product design helps to address common automated issues like: fissure cracks, bowl feeder jam, over/under torqued caps and cross threading.
Volume range of 0.5, 1.5 and 2.0mL.
Tube Color choices are an ultra-high clarity, natural color or amber option which provides ultra violet (UV) protection of 1.22% transmittance between 220 and 500nm.
Standard or Tethered cap, all available in a rainbow of colors. Visit cap product page for additional information.
Knurl tubes feature the ability to lock tube into specialty racks for single-hand operation.
Non knurl tubes ideal for automation and labeling, tubes are plain, removed of any labeling interference.
Skirted or Self Standing tubes can stand upright on their own, no rack required.
Conical tubes requires a rack to support them in the upright position.
Low nucleic acid and protein binding and low enzyme inhibition are features that Resin selection features.
RCF range from 18,000 to 25,000 x g.
Temperature Range -80°C to +95°C. Short-term storage in -80°C, -20°C, dry ice and wet ice. Short-term storage in vapor phase LN2. Suitable for a quick dip flash freeze in LN2.
Water Bath compatible up to +95°C.
† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.