Castor oil, specified according to the requirements of Ph.Eur.
Castor oil, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals

Castor oil, specified according to the requirements of Ph.Eur.

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 348855000
Price (USD)/ Each
74.65
Online exclusive
83.10 
Save 8.45 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
74.65
Online exclusive
83.10 
Save 8.45 (10%)
Add to cart
Castor oil, specified according to the requirements of Ph.Eur.
Catalog number348855000
Price (USD)/ Each
74.65
Online exclusive
83.10 
Save 8.45 (10%)
-
Add to cart
Chemical Identifiers
CAS8001-79-4
IUPAC Name1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
Molecular FormulaC57H104O9
InChI KeyZEMPKEQAKRGZGQ-UHFFFAOYNA-N
SMILESCCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.3 %
Angle of optical rotation+3.5° to +6.0° (20°C, 589 nm) (neat)
Absorbance=<0.7 (270 nm, 1 % in EtOH 96%)
Acid value=<1.5 mg KOH/g
Hydroxyl value>=160 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only