Unfortunately, due to limitations of our suppliers, we will no longer be able to offer this product after November 1, 2016 or when stocks run out, whichever occurs first.

Dichloromethane, HPLC Grade 4L

Catalog number: 400142

 Related applications: Protein Sequencing & Peptide Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Dichloromethane for use with Autoanalysis Trityl Monitor on ABI 392, 394

For Research Use Only. Not for use in diagnostics procedures.


Form: Liquid
Product Size: 4 L
Reagent Type: Oligo Synthesis Reagent
Chemical Name: Dichloromethane
Shipping Condition: Room Temperature


Manuals & protocols