xMAP™ Sheath Fluid (20L)

Catalog number: 4050000

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

xMAP® Sheath Fluid is the delivery medium, which carries the sample to the optics component of the Luminex xMAP® technology based instruments.
The Luminex® 200™ System is sold separately.
For In Vitro Diagnostic Use.

Specifications

For Use With (Equipment): FlexMAP® 3D™ System, Luminex® Instruments
Product Line: Luminex™, xMAP™
Product Size: 20 L
Shipping Condition: Room Temperature

Contents & storage

Store at Room Temperature.

Documents

Manuals & protocols

Recommended products