MicroAmp™ Multi Removal Tool
Applied Biosystems™

MicroAmp™ Multi Removal Tool

The Applied Biosystems™ MicroAmp™ Multi-Removal Tool makes it easier to remove caps from tubes, microplates from thermal cyclers, and muchRead more
Catalog NumberQuantity
43139501 Tool
Catalog number 4313950
Price (JPY)
4,000
Online offer
Ends: 25-Dec-2026
6,700
Save 2,700 (40%)
Each
Quantity:
1 Tool
The Applied Biosystems™ MicroAmp™ Multi-Removal Tool makes it easier to remove caps from tubes, microplates from thermal cyclers, and much more. Use this multi-functional tool to open MicroAmp™ Tubes or GeneAmp™ Tubes with attached caps, remove microplates from a thermal cycler sample block, and remove strips of caps from reaction tubes in a microplate and Tray⁄Retainer assembly.

•
Multi-edged tool streamlines repetitive tasks
• Easy-to-use
tool makes frustrating steps effortless
•
Sturdy construction extends product life

Versatile and Ergonomic

The tool has multiple edges with design features that facilitate the separation of caps from tubes, microplates from thermal cyclers, tubes from thermal cyclers, and tubes from microplates. One edge contains a U-shaped notch to open tube caps. One edge has a ridged
tongue to remove a strip of capped tubes from a microplate. Two edges have prongs to remove microplates from thermal cyclers. Finally, two hook protrusions can insert into the short side
of a microplate and lift it from a sample block.

Quick and Easy

The MicroAmp™ Multi-Removal Tool speeds up the routine steps of a large experiment and reduces dropped plates and spilled samples. Take the irritation out of your workday.

Note

: See user's manual or package insert for limited label license and trademark information.
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Application)PCR
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Product TypeMulti Removal Tool
Quantity1 Tool
Shipping ConditionRoom Temperature
Product LineMicroAmp
Unit SizeEach
Contents & Storage
Contains 1 MicroAmp™ Multi Removal Tool. Store at room temperature.