Product Image
Applied Biosystems™

SNaPshot™ Multiplex Kit

The SNaPshot Multiplex Kit is part of the SNaPshot Multiplex System. The SNaPshot Multiplex System primer extension-based method enables multiplexingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4323151100 Reactions, with protocol
4323159100 Reactions
43231611000 Reactions
43231635000 Reactions
Catalog number 4323151
Price (USD)
783.65
Online exclusive
816.00
Save 32.35
Each
No. of Reactions:
100 Reactions, with protocol
Price (USD)
783.65
Online exclusive
816.00
Save 32.35
Each

The SNaPshot Multiplex Kit is part of the SNaPshot Multiplex System. The SNaPshot Multiplex System primer extension-based method enables multiplexing up to 10 single nucleotide polymorphisms (SNPs). Use this system to screen and confirm SNPs, detect minor sequence variations, assess DNA methylation, fingerprint bacterial artificial chromosomes (BACs) or general labeling of restriction fragments.

• Interrogate up to 10 SNPs from different amplicons in a single base extension reaction
• Separate SNP loci that differ by a single base pair
• Increased sensitivity compared to standard sequencing allows detection of minor variants (e.g. somatic mutations) in a test sample
• Reduce the complexity of your experiments with this easy to use and cost-efficient genotyping system
• A large body of scientifc literature demonstrates the utility for genotyping, mutation detection and mapping

The SNaPshot Multiplex Kit is a part of the SNaPshot Multiplex System:
1) SNaPshot Multiplex Kit (100, 1000, 5000 reaction sizes)
2) GeneScan-120 LIZ Size Standard
3) Matrix Standard Set DS-02 for dye set E5 appropriate for your instrument

Each of the three components above is required to run the SNaPshot Multiplex Kit. The Matrix Standard set, however, is only required at a one-time initial set up to calibrate the instrument for the SNaPshot-specific dye set E5.

4) GeneMapper Software v.4.0 (or higher version) is the recommended secondary data analysis software that enables set up of user-defined panels and bins for automated data analysis.

For details, please see the Genemapper software and SNaPshot kit analysis "Getting Started Guide" below.

Screen and Confirm SNPs
The SNaPshot Multiplex System is the perfect tool for SNP screening and validation. The kit offers a one-tube single-base extension⁄termination reagent to label DNA fragments. Run DNA fragments on any Applied Biosystems capillary electrophoresis instrument with the GeneScan-120 LIZ Size Standard to indicate the size of labeled fragments. Then use GeneMapper Software to analyze the data and generate allele calls. The SNaPshot Multiplex Kit allows multiplexing during single-base extension of up to 10 primer-template combinations in a single tube-single capillary format.

Assess DNA Methylation
The study of methylation⁄epigenetics is emerging as an important component of cancer research. In a typical assay to detect methylation, bisulfite treatment of DNA deaminates non-methylated cytosine and converts it to uracil while methylated cytosine remains unreactive. The subsequent step of PCR amplification coverts uracil bases to thymine. Use SNaPshot todetect the base differences in treated and untreated samples to learn the methylation status of your samples.

Fingerprint BACs
New BAC libraries require a rapid, efficient method for characterization (also called fingerprinting) and assembly of clones into contigs (contiguous consensus sequences), which are then arrayed into physical maps of the chromosome. Use the SNaPshot Multiplex Kit to fingerprint clones by labeling BAC fragments after digestion with restriction endonucleases. The labeled fragments can then be separated and detected on any Applied Biosystems capillary electrophoresis instrument. Sizing information from GeneMapper Software v 4.0 or higher is imported into subsequent editing and contig assembly programs.

In a similar fashion, any restriction fragment with a 5' overhang can be labeled with a fluorescent dye using the SNaPshot reagent.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer, 3730/3730xl Genetic Analyzers
FormatKit
No. of Reactions100 Reactions, with protocol
Product LineSNaPshot
Product TypeMultiplex Kit
Quantity100 reactions
Run Time9 hr.
Sample TypeDNA
SpeciesAll Species
TargetSNPs (Known)
TechniqueSNP Genotyping (Fragment Analysis-Based), Primer Extension
Detection MethodPrimer-probe
For Use With (Application)Genotyping
Reaction Speed9 hr.
Unit SizeEach
Contents & Storage
Contains:
• SNaPshot™ Multiplex Ready Reaction Mix (for 100 reactions)
• SNaPshot™ Multiplex Control Primer Mix (30 μl)
• SNaPshot™ Multiplex Control Template (60 μl)
• Protocol

Store at -15°C to -25°C in a constant-temperature freezer.

Frequently asked questions (FAQs)

How many SNPs can the SNaPshot Multiplex kit analyze?

The SNaPshot Multiplex Kis is designed to interrogate up to 10 single nucleotide polymorphisms (SNPs) at known locations on 1-10 templates in a single tube.